Methyl pivalate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl trimethylacetate

12795-1000 12795-2500
TS12795-1000EA 36 USD
TS12795-1000 TS12795-2500
Methyl pivalate 99%
Methyl pivalate
Formula: C₆H₁₂O₂
Boiling Pt: 101 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.87 g/cm³
Flash Pt: <1 °C
Storage Temperature: Ambient
MDL Number: MFCD00008843
CAS Number: 598-98-1
EINECS: 209-959-1
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3890 to 1.3910 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR