Methyl methanesulphonate 99%

Supplier: Thermo Scientific Chemicals
Danger

15689-0050 15689-0250 15689-1000
TS15689-0050EA 61 USD
TS15689-0050 TS15689-0250 TS15689-1000
Methyl methanesulphonate 99%
Methyl methanesulphonate
Formula: C₂H₆O₃S
Boiling Pt: 202…203 °C (760 mmHg)
Melting Pt: 20 °C
Density: 1.3 g/cm³
Flash Pt: 104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007557
CAS Number: 66-27-3
EINECS: 200-625-0
UN: 2810
ADR: 6.1,III
Merck Index: 12,06170

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4130 to 1.4150 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR