Methyl trifluoroacetate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trifluoroacetic methyl ester

24341-0250 24341-1000
TS24341-0250EA 97 USD
TS24341-0250 TS24341-1000
Methyl trifluoroacetate 99%
Methyl trifluoroacetate
Formula: C₃H₃F₃O₂
MW: 128.05 g/mol
Boiling Pt: 43…43.5 °C (760 mmHg)
Melting Pt: –78 °C
Density: 1.273 g/cm³
Flash Pt: –7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000417
CAS Number: 431-47-0
EINECS: 207-074-5
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR