Ethyl hexanoate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl caproate, Caproic acid ethyl ester, Hexanoic acid ethyl ester

16499-0050 16499-1000 16499-5000
TS16499-0050EA 29 USD
TS16499-0050 TS16499-1000 TS16499-5000
Ethyl hexanoate 99+%
Ethyl hexanoate
Formula: C₈H₁₆O₂
Boiling Pt: 168 °C (760 mmHg)
Melting Pt: 67 °C
Density: 0.869 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00009511
CAS Number: 123-66-0
EINECS: 204-640-3
UN: 3272
ADR: 3,III
Merck Index: 13,03812

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4090 (20°C, 589 nm)
Water ≤0.3 % (K.F.)
Density (20°C) 0.867 to 0.871 kg/l
GC ≥99.0 %
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR