Pyruvic acid 98%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid

13214-0025 13214-0250 13214-1000 13214-5000
TS13214-0025EA 546 USD
TS13214-0025 TS13214-0250 TS13214-1000 TS13214-5000
Pyruvic acid 98%, Extra Pure
Pyruvic acid
Formula: C₃H₄O₃
MW: 88.06 g/mol
Boiling Pt: 165 °C (760 mmHg)
Melting Pt: 11.8 °C
Density: 1.25 g/cm³
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002585
CAS Number: 127-17-3
EINECS: 204-824-3
UN: 3265
ADR: 8,II
Merck Index: 12,08205

Order Now

Specification Test Results

Appearance Clear colorless to yellow to amber Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4320 (20°C, 589 nm)
Specific gravity (20°C) 1.240 to 1.280
Titration with NaOH 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR