Methyl pyruvate 94%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyruvic acid ethyl mester, 2-Ketopropionic acid methyl ester, 2-Oxopropionic acid methyl ester, Methyl 2-oxopropanoate, α-Ketopropionic acid methyl ester

12765-0250 12765-1000 12765-5000
TS12765-0250EA 37 USD
TS12765-0250 TS12765-1000 TS12765-5000
Methyl pyruvate 94%
Methyl pyruvate
Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 134…137 °C (760 mmHg)
Density: 1.13 g/cm³
Flash Pt: 39 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008754
CAS Number: 600-22-6
EINECS: 209-987-4
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4035 to 1.4075 (20°C, 589 nm)
GC ≥92 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR