Ethyl pyruvate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-oxopropanoate, 2-Oxopropionic acid ethyl ester, α-Ketopropionic acid ethyl ester, Pyruvic acid ethyl ester, 2-Ketopropionic acid ethyl ester

11894-1000 11894-5000
TS11894-1000EA 98 USD
TS11894-1000 TS11894-5000
Ethyl pyruvate 98%
Ethyl pyruvate
Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 144…147 °C (1013 hPa)
Melting Pt: –58 °C
Density: 1.0453 g/cm³
Flash Pt: 45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009123
CAS Number: 617-35-6
EINECS: 210-511-2
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4060 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR