3,4-Dimethoxycinnamic acid predominantly trans 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-Dimethoxycinnamic Acid

11551-0050 11551-0250 11551-1000
TS11551-0050EA 22 USD
TS11551-0050 TS11551-0250 TS11551-1000
3,4-Dimethoxycinnamic acid predominantly trans 99%
3,4-Dimethoxycinnamic acid
Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 181…184 °C
MDL Number: MFCD00004387
CAS Number: 2316-26-9
EINECS: 219-025-5

Order Now

Specification Test Results

Appearance Light yellow to light beige Powder
Infrared spectrum Conforms
Melting point 181 to 184 °C
GC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR