Methyl propionate 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Propanoic acid methyl ester

14905-0010 14905-2500
TS14905-0010EA 177 USD
TS14905-0010 TS14905-2500
Methyl propionate 99+%
Methyl propionate
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 79.7 °C (1013 hPa)
Melting Pt: –88 °C
Density: 0.915 g/cm³
Flash Pt: –2 °C
Storage Temperature: Ambient
MDL Number: MFCD00009306
CAS Number: 554-12-1
EINECS: 209-060-4
UN: 1248
ADR: 3,II
Merck Index: 12,06191

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3750 to 1.3790 (20°C, 589 nm)
Water ≤0.3 % (K.F.)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR