Ethyl propionate 99+%

Supplier: Thermo Scientific Chemicals
Danger

15050-0010 15050-0025 15050-0250 15050-5000
TS15050-0010EA 69 USD
TS15050-0010 TS15050-0025 TS15050-0250 TS15050-5000
Ethyl propionate 99+%
Ethyl propionate
Formula: C₅H₁₀O₂
Boiling Pt: 99 °C (760 mmHg)
Melting Pt: –73 °C
Density: 0.887 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00009308
CAS Number: 105-37-3
EINECS: 203-291-4
UN: 1195
ADR: 3,II
Merck Index: 13,03878

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3830 to 1.3870 (20°C, 589 nm)
GC ≥99.0 %
Relative density (20/4°C) 0.888 to 0.891
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR