Butyl propionate 99+%

Supplier: Thermo Scientific Chemicals
Warning

26948-0010 26948-2500
TS26948-0010EA 153 USD
TS26948-0010 TS26948-2500
Butyl propionate 99+%
Butyl propionate
Formula: C₇H₁₄O₂
Boiling Pt: 145 °C (760 mmHg)
Melting Pt: –75 °C
Density: 0.88 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00009448
CAS Number: 590-01-2
EINECS: 209-669-5
UN: 1914
ADR: 3,III
Merck Index: 12,01623

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4000 to 1.4020 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR