Ethyl valerate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl Pentanoate, Valeric acid ethyl ester

41047-0050 41047-1000 41047-5000
TS41047-0050EA 32 USD
TS41047-0050 TS41047-1000 TS41047-5000
Ethyl valerate 99%
Ethyl valerate
Formula: C₇H₁₄O₂
Boiling Pt: 145 °C
Melting Pt: –91 °C
Density: 0.87 g/cm³
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00009479
CAS Number: 539-82-2
EINECS: 208-726-1
UN: 3272
ADR: 3,III
Merck Index: 12,10042

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3990 to 1.4010 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR