Triethyl phosphate 99%

Supplier: Thermo Scientific Chemicals
Warning

13968-0010 13968-0025
TS13968-0010EA 118 USD
TS13968-0010 TS13968-0025
Triethyl phosphate 99%
Triethyl phosphate
Formula: C₆H₁₅O₄P
MW: 182.16 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: –56 °C
Density: 1.07 g/cm³
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00009077
CAS Number: 78-40-0
EINECS: 201-114-5
Merck Index: 12,09806

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4070 (20°C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %
Color scale ≤20 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR