Ethyl octanoate 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Caprylic acid ethyl ester, Octanoic acid ethyl ester, Ethyl caprylate

15051-0050 15051-1000 15051-5000
TS15051-0050EA 20 USD
TS15051-0050 TS15051-1000 TS15051-5000
Ethyl octanoate 99+%
Ethyl octanoate
Formula: C₁₀H₂₀O₂
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: –47 °C
Density: 0.868 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00009552
CAS Number: 106-32-1
EINECS: 203-385-5
Merck Index: 13,03813

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4200 (20°C, 589 nm)
Water ≤0.3 % (K.F.)
GC ≥99.0 %
Specific gravity (20°C) 0.866 to 0.87
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR