Dimethyl oxalate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Oxalic acid dimethyl ester

15927-1000 15927-5000
TS15927-1000EA 48 USD
TS15927-1000 TS15927-5000
Dimethyl oxalate 99%
Dimethyl oxalate
Formula: C₄H₆O₄
MW: 118.09 g/mol
Boiling Pt: 163 °C (1013 hPa)
Melting Pt: 51…55 °C
Density: 1.14 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00008442
CAS Number: 553-90-2
EINECS: 209-053-6
Merck Index: 12,06181

Order Now

Specification Test Results

Appearance Colorless to white Crystalline solid
Infrared spectrum Conforms
Melting point 51 to 55 °C
GC ≥98.5 %
UV at 280 nm A: ≤0.05
Titration ≥99.0 %
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR