Dimethyl maleate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethyl Maleate, Maleic acid dimethyl ester

21846-0010 21846-0051 21846-2500
TS21846-0010EA 182 USD
TS21846-0010 TS21846-0051 TS21846-2500
Dimethyl maleate 96%
Dimethyl maleate
Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 205
Melting Pt: -17.5
Density: 1.15 g/cm³
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00008459
CAS Number: 624-48-6
EINECS: 210-848-5

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4430 (20°C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥95 %
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR