4-Methoxyphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

30948-0010 30948-0050 30948-0250
TS30948-0010EA 41 USD
TS30948-0010 TS30948-0050 TS30948-0250
4-Methoxyphenylboronic acid 97%
4-Methoxyphenylboronic acid
Formula: C₇H₉BO₃
Melting Pt: 208…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00039139
CAS Number: 5720-07-0

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR