4-(Trifluoromethoxy)phenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dihydroxy(4-(trifluoromethoxy)phenyl)borane, (4-(Trifluoromethoxy)phenyl)metaboric acid, 4-(Trifluoromethoxy)benzeneboronic acid, 4-Trifluoromethoxyphenylboronic acid

35973-0010 35973-0050
TS35973-0010EA 51 USD
TS35973-0010 TS35973-0050
4-(Trifluoromethoxy)phenylboronic acid 98%
4-(Trifluoromethoxy)phenylboronic acid
Formula: C₇H₆BF₃O₃
MW: 205.93 g/mol
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD01074648
CAS Number: 139301-27-2

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR