Isovaleric acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Methylbutyric acid, Isopropyl acetic acid, 3-Methylbutanoic acid

15669-0025 15669-0100 15669-1000 15669-5000
TS15669-0025EA 262 USD
TS15669-0025 TS15669-0100 TS15669-1000 TS15669-5000
Isovaleric acid 99%
Isovaleric acid
Formula: C₅H₁₀O₂
Boiling Pt: 176 °C (760 mmHg)
Melting Pt: –35 °C
Density: 0.926 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00002726
CAS Number: 503-74-2
EINECS: 207-975-3
UN: 3265
ADR: 8,II
Merck Index: 12,05251

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3990 to 1.4070 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (25°C) 0.921 to 0.93


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR