3-Methoxyphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

35957-0010 35957-0050 35957-0100
TS35957-0010EA 35 USD
TS35957-0010 TS35957-0050 TS35957-0100
3-Methoxyphenylboronic acid 97%
3-Methoxyphenylboronic acid
Formula: C₇H₉BO₃
Melting Pt: 149…154 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00161359
CAS Number: 10365-98-7

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR