Methyl-3-mercaptopropionate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Mercaptopropionic acid methyl ester, Methyl 3-mercaptopropanoate

14887-0050 14887-1000 14887-5000
TS14887-0050EA 28 USD
TS14887-0050 TS14887-1000 TS14887-5000
Methyl-3-mercaptopropionate 98%
Methyl-3-mercaptopropionate
Formula: C₄H₈O₂S
MW: 120.17 g/mol
Boiling Pt: 54 °C (1013 hPa)
Melting Pt: –60 °C
Density: 1.08 g/cm³
Flash Pt: 60 °C
MDL Number: MFCD00004895
CAS Number: 2935-90-2
EINECS: 220-912-4
UN: 3071
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4640 (20°C, 589 nm)
Assay ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR