3,4-Dimethoxyphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

36939-0010 36939-0050 36939-0250
TS36939-0010EA 25 USD
TS36939-0010 TS36939-0050 TS36939-0250
3,4-Dimethoxyphenylboronic acid 97%
3,4-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
Melting Pt: 238…240 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074574
CAS Number: 122775-35-3

Order Now

Specification Test Results

Appearance White to beige Powder or granules and/or chunks
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR