3,4-Dichlorophenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
35909-0050
35909-0250
TS35909-0050EA
102
USD
TS35909-0050
TS35909-0250
3,4-Dichlorophenylboronic acid 97%
3,4-Dichlorophenylboronic acid
Formula:
C₆H₅BCl₂O₂ Melting Pt: 284…286 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01074646 CAS Number: 151169-75-4 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...