3,3-Dimethylbutyric acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,3-Dimethylbutanoic acid, tert-Butylacetic acid

10777-0100 10777-0500 10777-2500
TS10777-0100EA 30 USD
TS10777-0100 TS10777-0500 TS10777-2500
3,3-Dimethylbutyric acid 98%
3,3-Dimethylbutyric acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 185…190 °C (760 mmHg)
Melting Pt: 8…6 °C
Density: 0.91 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00002715
CAS Number: 1070-83-3
EINECS: 213-982-2
UN: 3265
ADR: 8,III
Merck Index: 13,01537

Order Now

Specification Test Results

Appearance Yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4090 to 1.4110 (20°C, 589 nm)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR