Isobutyric acid 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methylpropionic acid, 2-Methylpropanoic acid
12252-0010
12252-0250
12252-5000
TS12252-0010EA
76
USD
TS12252-0010
TS12252-0250
TS12252-5000
Isobutyric acid 99+%
Isobutyric acid
Formula:
C₄H₈O₂ MW: 88.11 g/mol Boiling Pt: 155 Melting Pt: -45 Density: 0.95 g/cm³ Flash Pt: 59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002658 CAS Number: 79-31-2 EINECS: 201-195-7 UN: 2529 ADR: 3 III Merck Index: 13,05172 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3920 to 1.3940 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...