2,4-Dichlorophenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dihydroxy(2,4-dichlorophenyl)borane, 2,4-Dichlorobenzeneboronic acid, (2,4-Dichlorophenyl)metaboric acid
36767-0010
36767-0050
TS36767-0010EA
23
USD
TS36767-0010
TS36767-0050
2,4-Dichlorophenylboronic acid 97%
2,4-Dichlorophenylboronic acid
Formula:
C₆H₅BCl₂O₂ MW: 190.82 g/mol Melting Pt: 240…245 °C |
MDL Number:
MFCD00013930 CAS Number: 68716-47-2 |
Specification Test Results
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...