Succinic acid ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Butanedioic acid

21955-0010 21955-2500
TS21955-0010EA 123 USD
TS21955-0010 TS21955-2500
Succinic acid ACS
Succinic acid
Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235 °C (760 mmHg)
Melting Pt: 185…190 °C
Flash Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
Merck Index: 13,08953
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd381e6e0e4cbde676a92c9e8d6aacfac635ade&vid=0b14e1b7f82b03468fc82d224a0ee894952eb200&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Melting point 185.0 to 191.0°C
Sulfated ash ≤0.02 %
Titration with NaOH ≥99.0 %
Chloride (Cl) ≤0.001 %
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.003 %
Iron (Fe) ≤0.0005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 %
Nitrogen compounds (as N) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR