(S)-(-)-2-Chloropropionic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-(-)-2-Chloropropanoic acid

25497-0010 25497-0050
TS25497-0010EA 42 USD
TS25497-0010 TS25497-0050
(S)-(-)-2-Chloropropionic acid 98%
(S)-(-)-2-Chloropropionic acid
Formula: H₃CCHClCOOH
MW: 108.52 g/mol
Boiling Pt: 187 °C (760 mmHg)
Melting Pt: 4 °C
Density: 1.24 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00064205
CAS Number: 29617-66-1
EINECS: 411-150-5
UN: 2511
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4360 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation -14° to -15° (25°C, 589 nm) (neat)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR