(S)-(-)-2-Bromopropionic acid 98%, >85% ee

Supplier: Thermo Scientific Chemicals
Danger

27791-0050 27791-0250 27791-1000
TS27791-0050EA 72 USD
TS27791-0050 TS27791-0250 TS27791-1000
(S)-(-)-2-Bromopropionic acid 98%, >85% ee
(S)-(-)-2-Bromopropionic acid
Formula: H₃CBrCHCOOH
Boiling Pt: 203 °C (760 mmHg)
Melting Pt: –35 °C
Density: 1.7 g/cm³
Flash Pt: 100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00137325
CAS Number: 32644-15-8
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4760 (20°C, 589 nm)
HPLC ≥97.5 %
Enantiomeric excess ≥80 % (Typically)
Specific optical rotation -24° to -30° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR