(S)-(+)-Hydratropic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-2-Phenylpropanoic acid, (S)-(+)-2-Phenylpropionic acid, (S)-(+)-2-Phenylpropanoic acid
26465-0010
26465-2500
TS26465-0010EA
194
USD
TS26465-0010
TS26465-2500
(S)-(+)-Hydratropic acid 99%
(S)-(+)-Hydratropic acid
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 115 °C (1 mmHg) Melting Pt: 29…32 °C Density: 1.1 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063139 CAS Number: 7782-24-3 |
Specification Test Results
Appearance | Off-white Low melting solid |
Infrared spectrum | Conforms |
Melting point | 29 to 32 °C |
GC | ≥98.5 % |
Specific optical rotation | +74° to +76° (20 °C, 589 nm) (c=3, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...