L(+)-Lactic acid 20% (w/w) in water
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid
12507-0010
12507-0025
TS12507-0010EA
87
USD
TS12507-0010
TS12507-0025
L(+)-Lactic acid 20% (w/w) in water
L(+)-Lactic acid
Formula:
C₃H₆O₃ MW: 90.08 g/mol Density: 1.05 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00064266 CAS Number: 79-33-4 EINECS: 201-196-2 |
Specification Test Results
Appearance | Clear colorless Solution |
Titration with NaOH | ≥18 % |
Sulfate (SO4) | ≤30 ppm |
Iron (Fe) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...