(4-Biphenyl)acetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Biphenylacetic acid
18815-0100
18815-0250
TS18815-0100EA
55
USD
TS18815-0100
TS18815-0250
(4-Biphenyl)acetic acid 98%
(4-Biphenyl)acetic acid
Formula:
(C₆H₅C₆H₅)CH₂COOH Boiling Pt: 365 °C (1013 hPa) Melting Pt: 156…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004351 CAS Number: 5728-52-9 EINECS: 227-233-2 UN: 2811 ADR: 6.1,III Merck Index: 13,03979 |
Specification Test Results
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Melting point | 158 to 165 °C |
Titration with TBAH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...