1,2-Diphenylethane 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Dibenzyl, Bibenzyl

10623-0050 10623-0250 10623-1000
TS10623-0050EA 26 USD
TS10623-0050 TS10623-0250 TS10623-1000
1,2-Diphenylethane 99%
1,2-Diphenylethane
Formula: (C₆H₅)CH₂CH₂(C₆H₅)
MW: 182.27 g/mol
Boiling Pt: 284 °C (760 mmHg)
Melting Pt: 49…53 °C
Density: 1.01 g/cm³
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD00004796
CAS Number: 103-29-7
EINECS: 203-096-4
Merck Index: 12,01245

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 49 to 53 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR