2,3-Dimethyl-2,3-diphenylbutane 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
1,1'-(1,1,2,2-Tetramethylethylene)dibenzene, (2,3-Dimethylbutane-2,3-diyl)dibenzene
34998-0250
34998-1000
TS34998-0250EA
34
USD
TS34998-0250
TS34998-1000
2,3-Dimethyl-2,3-diphenylbutane 95%
2,3-Dimethyl-2,3-diphenylbutane
Formula:
H₃CC(CH₃)(C₆H₅)C(CH₃)(C₆H₅)CH₃ MW: 238.37 g/mol Boiling Pt: 170…171 °C (14 mmHg) Melting Pt: 90…110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00053713 CAS Number: 1889-67-4 EINECS: 217-568-2 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or flakes and/or chunks |
Infrared spectrum | Conforms |
Assay | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...