(2,6-Dichlorophenyl)acetic acid methyl ester 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-(2,6-dichlorophenyl)acetate, Methyl 2,6-Dichlorophenylacetate, 2,6-Dichlorophenylacetic acid methyll ester, Guanfacine Methyl Ester Impurity, Methyl (2,6-dichlorophenyl)acetate

35107-0050 35107-0250
TS35107-0050EA 61 USD
TS35107-0050 TS35107-0250
(2,6-Dichlorophenyl)acetic acid methyl ester 99%
(2,6-Dichlorophenyl)acetic acid methyl ester
Formula: (C₆H₃Cl₂)CH₂COOCH₃
MW: 219.07 g/mol
MDL Number: MFCD00191640
CAS Number: 54551-83-6
EINECS: 259-221-8

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5420 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR