(±)-1-Penten-3-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-Ethyl vinyl carbinol, Pent-1-en-3-ol

13001-0050 13001-0250 13001-1000
TS13001-0050EA 26 USD
TS13001-0050 TS13001-0250 TS13001-1000
(±)-1-Penten-3-ol 97%
(±)-1-Penten-3-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 114…115 °C (760 mmHg)
Density: 0.839 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00004573
CAS Number: 616-25-1
EINECS: 210-472-1
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4260 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR