Diethyl methylphosphonate 96%

Supplier: Thermo Scientific Chemicals
Warning

33368-0050 33368-0250
TS33368-0050EA 73 USD
TS33368-0050 TS33368-0250
Diethyl methylphosphonate 96%
Diethyl methylphosphonate
Formula: C₅H₁₃O₃P
MW: 152.13 g/mol
Boiling Pt: 78…81 °C (15 mmHg)
Flash Pt: 75 °C
MDL Number: MFCD00009813
CAS Number: 683-08-9
EINECS: 211-667-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4130 to 1.4150 (20°C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR