Trifluoroacetamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2,2-Trifluoroacetamide
38522-0250
38522-2500
TS38522-0250EA
38
USD
TS38522-0250
TS38522-2500
Trifluoroacetamide 97%
Trifluoroacetamide
Formula:
C₂H₂F₃NO MW: 113.04 g/mol Boiling Pt: 162…164 °C (760 mmHg) Melting Pt: 59…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008008 CAS Number: 354-38-1 EINECS: 206-559-9 |
Specification Test Results
Appearance | White to light yellow to beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 68 to 75 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...