3,3-Dimethylglutaric acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,3-Dimethylpentanedioic acid, 3,3-Dimethyl-1,5-pentanedioic acid

11624-0250 11624-1000 11624-5000
TS11624-0250EA 72 USD
TS11624-0250 TS11624-1000 TS11624-5000
3,3-Dimethylglutaric acid 98%
3,3-Dimethylglutaric acid
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 290 °C (760 mmHg)
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00002716
CAS Number: 4839-46-7
EINECS: 225-425-0

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 102 °C
Water ≤1 % (K.F.)
Titration with NaOH ≥97.5 %
Phosphate (PO4) ≤2 ppm
Sulfate (SO4) ≤2 ppm
Iron (Fe) ≤0.2 ppm
Heavy metals (as Pb) ≤0.5 ppm
Carbon dioxide (CO2) ≤0.002 %
KMnO4-reducing substance Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR