2-Naphthylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Naphthaleneacetic acid, 2-(Naphthalen-2-yl)acetic acid

12822-0050 12822-0250 12822-1000
TS12822-0050EA 58 USD
TS12822-0050 TS12822-0250 TS12822-1000
2-Naphthylacetic acid 99%
2-Naphthylacetic acid
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Melting Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00004126
CAS Number: 581-96-4
EINECS: 209-475-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 141 to 143 °C
GC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR