Nicotinic acid (Niacin), meets analytical specification of Ph.Eur.

Supplier: Thermo Scientific Chemicals
Warning

38032-0025 38032-5000
TS38032-0025EA 818 USD
TS38032-0025 TS38032-5000
Nicotinic acid (Niacin), meets analytical specification of Ph.Eur.
Nicotinic acid (Niacin)
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Melting Pt: 236…239 °C
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 13,06552

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 234 to 240 °C
Loss on drying ≤1.0 %
Sulfated ash ≤0.1 %
Solubility (in water) Sparingly soluble
Related substances Passes test
Titration with NaOH 99.5 to 100.5 % (On dried substance)
Chloride (Cl) ≤200 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR