Diethyl maleate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Maleic acid diethyl ester, Diethyl (Z)-2-butenedioate

11444-0010 11444-0050 11444-2500
TS11444-0010EA 77 USD
TS11444-0010 TS11444-0050 TS11444-2500
Diethyl maleate 97%
Diethyl maleate
Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 225
Melting Pt: -11
Density: 1.06 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009191
CAS Number: 141-05-9
EINECS: 205-451-9
Merck Index: 13,03150

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4420 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR