Ethyl-3-mercaptopropionate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 3-mercaptopropanoate, 3-Mercaptopropionic acid ethyl ester

13741-0050 13741-0250
TS13741-0050EA 52 USD
TS13741-0050 TS13741-0250
Ethyl-3-mercaptopropionate 98%
Ethyl-3-mercaptopropionate
Formula: C₅H₁₀O₂S
MW: 134.2 g/mol
Boiling Pt: 75…76 °C (10 mmHg)
Density: 1.059 g/cm³
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004896
CAS Number: 5466-06-8
EINECS: 226-771-5
UN: 3334

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4560 to 1.4580 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR