2-Butynoic acid methyl ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-butynoate, Methyl but-2-ynoate

22349-0010 22349-0050 22349-0250
TS22349-0010EA 72 USD
TS22349-0010 TS22349-0050 TS22349-0250
2-Butynoic acid methyl ester 97%
2-Butynoic acid methyl ester
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 76
Density: 0.98 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00009273
CAS Number: 23326-27-4
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4360 to 1.4380 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR