Ethyl butyrate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Butanoic acid ethyl ester, Butyric acid ethyl ester

11818-0010 11818-0050 11818-2500
TS11818-0010EA 75 USD
TS11818-0010 TS11818-0050 TS11818-2500
Ethyl butyrate 99%
Ethyl butyrate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: –93.3 °C
Density: 0.87 g/cm³
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009394
CAS Number: 105-54-4
EINECS: 203-306-4
UN: 1180
ADR: 3,III
Merck Index: 13,03810

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3910 to 1.3930 (20°C, 589 nm)
Water ≤0.3 % (K.F.)
GC ≥98.5 %
Acid value ≤1 mg KOH/g
Potassium (K) ≤50 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR