2,3-Dichlorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

36771-0010 36771-0050 36771-0250
TS36771-0010EA 29 USD
TS36771-0010 TS36771-0050 TS36771-0250
2,3-Dichlorophenylboronic acid 97%
2,3-Dichlorophenylboronic acid
Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Melting Pt: 242…244 °C
Storage Temperature: Ambient
MDL Number: MFCD01075703
CAS Number: 151169-74-3

Order Now

Specification Test Results

Appearance White to cream Powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR