3-Nitrophenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(3-Nitrophenyl)metaboric acid, Dihydroxy(3-nitrophenyl)borane, 3-Nitrobenzeneboronic acid
34470-0010
34470-0050
TS34470-0010EA
28
USD
TS34470-0010
TS34470-0050
3-Nitrophenylboronic acid 97%
3-Nitrophenylboronic acid
Formula:
C₆H₆BNO₄ Melting Pt: 289…291 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007193 CAS Number: 13331-27-6 |
Specification Test Results
Appearance | Light yellow to light brown Powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...