1,2,3,4-Tetramethylcyclopentadiene mixture of isomers 85%
Supplier: Thermo Scientific Chemicals
|
Warning
|
34807-0050
34807-0250
TS34807-0050EA
156
USD
TS34807-0050
TS34807-0250
1,2,3,4-Tetramethylcyclopentadiene mixture of isomers 85%
1,2,3,4-Tetramethylcyclopentadiene
Formula:
C₉H₁₄ Boiling Pt: 142 °C Density: 0.808 g/cm³ Flash Pt: 42 °C Storage Temperature: Freezer |
MDL Number:
MFCD00145385 CAS Number: 4249-10-9 UN: 3295 ADR: 3,III |
Specification Test Results
Appearance | Colorless to yellow to orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4700 to 1.4760 (20°C, 589 nm) |
GC | ≥80 % (mixture of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...