Phloroglucinol dihydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3,5-Trihydroxybenzene dihydrate, Phloroglucinol, Hydrate

13104-0250 13104-1000 13104-5000
TS13104-0250EA 32 USD
TS13104-0250 TS13104-1000 TS13104-5000
Phloroglucinol dihydrate 99%
Phloroglucinol dihydrate
Formula: C₆H₆O₃·2H₂O
MW: 162.14 g/mol
Melting Pt: 218…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00149090
CAS Number: 6099-90-7
EINECS: 203-611-2
Merck Index: 13,07413

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR