(±)-4-Penten-2-ol 96%

Supplier: Thermo Scientific Chemicals
Warning

38743-0050 38743-0250
TS38743-0050EA 74 USD
TS38743-0050 TS38743-0250
(±)-4-Penten-2-ol 96%
(±)-4-Penten-2-ol
Formula: C₅H₁₀O
Boiling Pt: 115 °C (1013 hPa)
Density: 0.837 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00004556
CAS Number: 625-31-0
EINECS: 210-887-8
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4250 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR